3-Piperidinyl-2-methoxyacetate hydrochloride

Supplier: Matrix Scientific

049117-500MG 049117-2.5G
101886-372EA 364.62 USD
101886-372 101886-374
3-Piperidinyl-2-methoxyacetate hydrochloride
3-Piperidinyl-2-methoxyacetate hydrochloride

Hazard: Irritant

Formula: C₈H₁₆ClNO₃
MW: 209.67 g/mol
MDL Number: MFCD13559680
CAS Number: 1220031-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR