N-(2-Furoyl)-2-methylalanine

Supplier: Matrix Scientific

049115-500MG 049115-2.5G
101886-364EA 364.62 USD
101886-364 101886-366
N-(2-Furoyl)-2-methylalanine
N-(2-Furoyl)-2-methylalanine

Hazard: Irritant

Formula: C₉H₁₁NO₄
MDL Number: MFCD13559679
CAS Number: 1219979-72-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR