1-(2-Furoyl)-3-piperidinecarboxylic acid
Supplier: Matrix Scientific
049113-500MG
049113-2.5G
101886-356EA
364.62
USD
101886-356
101886-358
1-(2-Furoyl)-3-piperidinecarboxylic acid
1-(2-Furoyl)-3-piperidinecarboxylic acid
Hazard: Irritant
Formula:
C₁₁H₁₃NO₄ |
MDL Number:
MFCD02678422 CAS Number: 436093-09-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...