N-(4-Piperidinyl)-2-furamide hydrochloride
Supplier: Matrix Scientific
049110-500MG
049110-2.5G
101886-344EA
364.62
USD
101886-344
101886-346
N-(4-Piperidinyl)-2-furamide hydrochloride
N-(4-Piperidinyl)-2-furamide hydrochloride
Hazard: Irritant
Formula:
C₁₀H₁₅ClN₂O₂ MW: 230.7 g/mol |
MDL Number:
MFCD13559678 CAS Number: 1220037-32-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...