Methyl (2S,4S)-4-(2-furoyloxy)-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

049109-500MG 049109-2.5G
101886-340EA 364.62 USD
101886-340 101886-342
Methyl (2S,4S)-4-(2-furoyloxy)-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-(2-furoyloxy)-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₄ClNO₅
MDL Number: MFCD13559677
CAS Number: 1354487-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR