3-Pyrrolidinyl-2-furoate hydrochloride

Supplier: Matrix Scientific

049108-500MG 049108-2.5G
101886-336EA 364.62 USD
101886-336 101886-338
3-Pyrrolidinyl-2-furoate hydrochloride
3-Pyrrolidinyl-2-furoate hydrochloride

Hazard: Irritant

Formula: C₉H₁₂ClNO₃
MW: 217.65 g/mol
MDL Number: MFCD13559676
CAS Number: 1220031-53-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR