3-Pyrrolidinylmethyl-2-furoate hydrochloride

Supplier: Matrix Scientific

049107-500MG 049107-2.5G
101886-332EA 364.62 USD
101886-332 101886-334
3-Pyrrolidinylmethyl-2-furoate hydrochloride
3-Pyrrolidinylmethyl-2-furoate hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₄ClNO₃
MDL Number: MFCD13559675
CAS Number: 1220037-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR