2-(2-Piperidinyl)ethyl-2-furoate hydrochloride

Supplier: Matrix Scientific

049106-500MG 049106-2.5G
101886-328EA 364.62 USD
101886-328 101886-330
2-(2-Piperidinyl)ethyl-2-furoate hydrochloride
2-(2-Piperidinyl)ethyl-2-furoate hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₈ClNO₃
MDL Number: MFCD13559674
CAS Number: 1219949-55-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR