2-(4-Piperidinyl)ethyl-2-furoate hydrochloride
Supplier: Matrix Scientific
049104-500MG
049104-2.5G
101886-320EA
364.62
USD
101886-320
101886-322
2-(4-Piperidinyl)ethyl-2-furoate hydrochloride
2-(4-Piperidinyl)ethyl-2-furoate hydrochloride
Hazard: Irritant
Formula:
C₁₂H₁₈ClNO₃ |
MDL Number:
MFCD13559672 CAS Number: 1220031-55-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...