3-Piperidinylmethyl-2-furoate hydrochloride

Supplier: Matrix Scientific

049102-500MG 049102-2.5G
101886-312EA 364.62 USD
101886-312 101886-314
3-Piperidinylmethyl-2-furoate hydrochloride
3-Piperidinylmethyl-2-furoate hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₆ClNO₃
MW: 245.71 g/mol
MDL Number: MFCD13559670
CAS Number: 1220020-31-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR