N-[(4-Chloro-2-pyridinyl)carbonyl]-2-methylalanine

Supplier: Matrix Scientific

049096-500MG 049096-2.5G
101886-288EA 364.62 USD
101886-288 101886-290
N-[(4-Chloro-2-pyridinyl)carbonyl]-2-methylalanine
N-[(4-Chloro-2-pyridinyl)carbonyl]-2-methylalanine

Hazard: Irritant

Formula: C₁₀H₁₁ClN₂O₃
MDL Number: MFCD13559666
CAS Number: 1220031-50-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR