1-[(4-Chloro-2-pyridinyl)carbonyl]-4-piperidinecarboxylic acid

Supplier: Matrix Scientific

049095-500MG 049095-2.5G
101886-284EA 364.62 USD
101886-284 101886-286
1-[(4-Chloro-2-pyridinyl)carbonyl]-4-piperidinecarboxylic acid
1-[(4-Chloro-2-pyridinyl)carbonyl]-4-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₃ClN₂O₃
MDL Number: MFCD12094724
CAS Number: 1153769-18-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR