1-[(4-Chloro-2-pyridinyl)carbonyl]-3-piperidinecarboxylic acid

Supplier: Matrix Scientific

049094-500MG 049094-2.5G
101886-280EA 364.62 USD
101886-280 101886-282
1-[(4-Chloro-2-pyridinyl)carbonyl]-3-piperidinecarboxylic acid
1-[(4-Chloro-2-pyridinyl)carbonyl]-3-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₃ClN₂O₃
MDL Number: MFCD12094729
CAS Number: 1154331-74-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR