N-[(4-Chloro-2-pyridinyl)carbonyl]alanine

Supplier: Matrix Scientific

049091-500MG 049091-2.5G
101886-268EA 364.62 USD
101886-268 101886-270
N-[(4-Chloro-2-pyridinyl)carbonyl]alanine
N-[(4-Chloro-2-pyridinyl)carbonyl]alanine

Hazard: Irritant

Formula: C₉H₉ClN₂O₃
MDL Number: MFCD12094727
CAS Number: 1308946-57-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR