4-Chloro-N-(4-piperidinyl)-2-pyridinecarboxamide hydrochloride
Supplier: Matrix Scientific
049089-500MG
049089-2.5G
101886-260EA
364.62
USD
101886-260
101886-262
4-Chloro-N-(4-piperidinyl)-2-pyridinecarboxamide hydrochloride
4-Chloro-N-(4-piperidinyl)-2-pyridinecarboxamide hydrochloride
Hazard: Irritant
Formula:
C₁₁H₁₅Cl₂N₃O |
MDL Number:
MFCD13559665 CAS Number: 1220020-96-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...