N-(4-Amino-2-methoxyphenyl)-4-chloro-2-pyridinecarboxamide

Supplier: Matrix Scientific

049088-500MG 049088-2.5G
101886-256EA 364.62 USD
101886-256 101886-258
N-(4-Amino-2-methoxyphenyl)-4-chloro-2-pyridinecarboxamide
N-(4-Amino-2-methoxyphenyl)-4-chloro-2-pyridinecarboxamide

Hazard: Irritant

Formula: C₁₃H₁₂ClN₃O₂
MDL Number: MFCD13559664
CAS Number: 1220031-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR