3-Pyrrolidinyl-4-chloro-2-pyridinecarboxylate hydrochloride

Supplier: Matrix Scientific

049086-500MG 049086-2.5G
101886-248EA 364.62 USD
101886-248 101886-250
3-Pyrrolidinyl-4-chloro-2-pyridinecarboxylate hydrochloride
3-Pyrrolidinyl-4-chloro-2-pyridinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₂Cl₂N₂O₂
MDL Number: MFCD13559662
CAS Number: 1219948-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR