3-Pyrrolidinylmethyl-4-chloro-2-pyridinecarboxylate hydrochloride

Supplier: Matrix Scientific

049085-500MG 049085-2.5G
101886-244EA 364.62 USD
101886-244 101886-246
3-Pyrrolidinylmethyl-4-chloro-2-pyridinecarboxylate hydrochloride
3-Pyrrolidinylmethyl-4-chloro-2-pyridinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₄Cl₂N₂O₂
MDL Number: MFCD13559661
CAS Number: 1220031-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR