2-(2-Piperidinyl)ethyl-4-chloro-2-pyridinecarboxylate hydrochloride

Supplier: Matrix Scientific

049084-500MG 049084-2.5G
101886-240EA 364.62 USD
101886-240 101886-242
2-(2-Piperidinyl)ethyl-4-chloro-2-pyridinecarboxylate hydrochloride
2-(2-Piperidinyl)ethyl-4-chloro-2-pyridinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈Cl₂N₂O₂
MDL Number: MFCD13559660
CAS Number: 1219949-53-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR