2-(4-Piperidinyl)ethyl-4-chloro-2-pyridinecarboxylate hydrochloride

Supplier: Matrix Scientific

049082-500MG 049082-2.5G
101886-232EA 364.62 USD
101886-232 101886-234
2-(4-Piperidinyl)ethyl-4-chloro-2-pyridinecarboxylate hydrochloride
2-(4-Piperidinyl)ethyl-4-chloro-2-pyridinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈Cl₂N₂O₂
MDL Number: MFCD13559658
CAS Number: 1220031-37-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR