3-Piperidinylmethyl-4-chloro-2-pyridinecarboxylate hydrochloride

Supplier: Matrix Scientific

049080-500MG 049080-2.5G
101886-224EA 364.62 USD
101886-224 101886-226
3-Piperidinylmethyl-4-chloro-2-pyridinecarboxylate hydrochloride
3-Piperidinylmethyl-4-chloro-2-pyridinecarboxylate hydrochloride
Formula: C₁₂H₁₆Cl₂N₂O₂
MDL Number: MFCD13559656
CAS Number: 1220037-09-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR