4-Piperidinyl-4-chloro-2-pyridinecarboxylate hydrochloride

Supplier: Matrix Scientific

049078-500MG 049078-2.5G
101886-216EA 364.62 USD
101886-216 101886-218
4-Piperidinyl-4-chloro-2-pyridinecarboxylate hydrochloride
4-Piperidinyl-4-chloro-2-pyridinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₄Cl₂N₂O₂
MW: 277.15 g/mol
MDL Number: MFCD13559654
CAS Number: 1219949-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR