3-Piperidinyl-4-chloro-2-pyridinecarboxylate hydrochloride

Supplier: Matrix Scientific

049077-500MG 049077-2.5G
101886-212EA 364.62 USD
101886-212 101886-214
3-Piperidinyl-4-chloro-2-pyridinecarboxylate hydrochloride
3-Piperidinyl-4-chloro-2-pyridinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₄Cl₂N₂O₂
MDL Number: MFCD13559653
CAS Number: 1219949-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR