N-Isonicotinoyl-β-alanine

Supplier: Matrix Scientific

049076-500MG 049076-2.5G
101886-208EA 364.62 USD
101886-208 101886-210
N-Isonicotinoyl-β-alanine
N-Isonicotinoyl-beta-alanine

Hazard: Irritant

Formula: C₉H₁₀N₂O₃
MW: 194.19 g/mol
MDL Number: MFCD00463120
CAS Number: 147218-38-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR