1-Isonicotinoyl-2-piperidinecarboxylic acid
Supplier: Matrix Scientific
049072-500MG
049072-2.5G
101886-192EA
364.62
USD
101886-192
101886-194
1-Isonicotinoyl-2-piperidinecarboxylic acid
1-Isonicotinoyl-2-piperidinecarboxylic acid
Hazard: Irritant
Formula:
C₁₂H₁₄N₂O₃ |
MDL Number:
MFCD08442848 CAS Number: 67691-58-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...