2-(Isonicotinoylamino)acetic acid

Supplier: Matrix Scientific

049069-500MG 049069-2.5G
101886-180EA 364.62 USD
101886-180 101886-182
2-(Isonicotinoylamino)acetic acid
2-(Isonicotinoylamino)acetic acid

Hazard: Irritant

Formula: C₈H₈N₂O₃
MDL Number: MFCD00995797
CAS Number: 2015-20-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR