N-(4-Piperidinyl)isonicotinamide hydrochloride

Supplier: Matrix Scientific

049068-500MG 049068-2.5G
101886-176EA 364.62 USD
101886-176 101886-178
N-(4-Piperidinyl)isonicotinamide hydrochloride
N-(4-Piperidinyl)isonicotinamide hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₆ClN₃O
MDL Number: MFCD13559651
CAS Number: 1219979-61-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR