3-Pyrrolidinyl isonicotinate hydrochloride

Supplier: Matrix Scientific

049066-500MG 049066-2.5G
101886-168EA 364.62 USD
101886-168 101886-170
3-Pyrrolidinyl isonicotinate hydrochloride
3-Pyrrolidinyl isonicotinate hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₃ClN₂O₂
MW: 228.68 g/mol
MDL Number: MFCD13559649
CAS Number: 1219979-35-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR