2-(3-Piperidinyl)ethyl isonicotinate hydrochloride

Supplier: Matrix Scientific

049063-500MG 049063-2.5G
101886-156EA 364.62 USD
101886-156 101886-158
2-(3-Piperidinyl)ethyl isonicotinate hydrochloride
2-(3-Piperidinyl)ethyl isonicotinate hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉ClN₂O₂
MDL Number: MFCD13559646
CAS Number: 1219979-49-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR