2-(4-Piperidinyl)ethyl isonicotinate hydrochloride

Supplier: Matrix Scientific

049062-500MG 049062-2.5G
101886-152EA 469.23 USD
101886-152 101886-154
2-(4-Piperidinyl)ethyl isonicotinate hydrochloride
2-(4-Piperidinyl)ethyl isonicotinate hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉ClN₂O₂
MDL Number: MFCD13559645
CAS Number: 1219979-31-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR