3-Piperidinylmethyl isonicotinate hydrochloride

Supplier: Matrix Scientific

049060-500MG 049060-2.5G
101886-144EA 364.62 USD
101886-144 101886-146
3-Piperidinylmethyl isonicotinate hydrochloride
3-Piperidinylmethyl isonicotinate hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂O₂
MW: 256.73 g/mol
MDL Number: MFCD13559643
CAS Number: 1220037-02-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR