3-Azetidinyl isonicotinate

Supplier: Matrix Scientific

049059-500MG 049059-2.5G
101886-140EA 364.62 USD
101886-140 101886-142
3-Azetidinyl isonicotinate
3-Azetidinyl isonicotinate

Hazard: Irritant

Formula: C₉H₁₀N₂O₂
MDL Number: MFCD13559642
CAS Number: 1219979-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR