3-Pyrrolidinyl acetate hydrochloride

Supplier: Matrix Scientific

049048-500MG 049048-2.5G
101886-096EA 364.62 USD
101886-096 101886-098
3-Pyrrolidinyl acetate hydrochloride
3-Pyrrolidinyl acetate hydrochloride

Hazard: Irritant

Formula: C₆H₁₂ClNO₂
MDL Number: MFCD13559637
CAS Number: 1219949-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR