2-(3-Piperidinyl)ethyl acetate hydrochloride

Supplier: Matrix Scientific

049045-500MG 049045-2.5G
101886-084EA 364.62 USD
101886-084 101886-086
2-(3-Piperidinyl)ethyl acetate hydrochloride
2-(3-Piperidinyl)ethyl acetate hydrochloride

Hazard: Irritant

Formula: C₉H₁₈ClNO₂
MDL Number: MFCD13559634
CAS Number: 1220031-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR