2-(4-Piperidinyl)ethyl acetate hydrochloride
Supplier: Matrix Scientific
049044-500MG
049044-2.5G
101886-080EA
186.15
USD
101886-080
101886-082
2-(4-Piperidinyl)ethyl acetate hydrochloride
2-(4-Piperidinyl)ethyl acetate hydrochloride
Hazard: Irritant
Formula:
C₉H₁₈ClNO₂ |
MDL Number:
MFCD13559633 CAS Number: 184042-52-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...