4-Piperidinylmethyl acetate hydrochloride

Supplier: Matrix Scientific

Synonyms: Piperidin-4-ylmethyl acetate hydrochloride

049043-500MG 049043-2.5G
101886-076EA 364.62 USD
101886-076 101886-078
4-Piperidinylmethyl acetate hydrochloride
4-Piperidinylmethyl acetate hydrochloride

Hazard: Irritant

Formula: C₈H₁₆ClNO₂
MW: 193.68 g/mol
MDL Number: MFCD13559632
CAS Number: 184042-50-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR