3-Piperidinylmethyl acetate hydrochloride

Supplier: Matrix Scientific

049042-500MG 049042-2.5G
101886-072EA 364.62 USD
101886-072 101886-074
3-Piperidinylmethyl acetate hydrochloride
3-Piperidinylmethyl acetate hydrochloride

Hazard: Irritant

Formula: C₈H₁₆ClNO₂
MW: 193.68 g/mol
MDL Number: MFCD13559631
CAS Number: 1219979-17-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR