3-Azetidinyl acetate

Supplier: Matrix Scientific

Synonyms: Azetidin-3-yl acetate

049041-500MG 049041-2.5G
101886-068EA 364.62 USD
101886-068 101886-070
3-Azetidinyl acetate
3-Azetidinyl acetate

Hazard: Irritant

Formula: C₅H₉NO₂
MW: 115.13 g/mol
MDL Number: MFCD13559630
CAS Number: 74121-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR