4-Piperidinyl acetate hydrochloride

Supplier: Matrix Scientific

049040-500MG 049040-2.5G
101886-064EA 364.62 USD
101886-064 101886-066
4-Piperidinyl acetate hydrochloride
4-Piperidinyl acetate hydrochloride

Hazard: Irritant

Formula: C₇H₁₄ClNO₂
MDL Number: MFCD08061428
CAS Number: 94886-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR