N-(4-Amino-2-chlorophenyl)-2-(3-fluorophenoxy)acetamide

Supplier: Matrix Scientific

049038-500MG 049038-2.5G
101886-056EA 364.62 USD
101886-056 101886-058
N-(4-Amino-2-chlorophenyl)-2-(3-fluorophenoxy)acetamide
N-(4-Amino-2-chlorophenyl)-2-(3-fluorophenoxy)acetamide

Hazard: Irritant

Formula: C₁₄H₁₂ClFN₂O₂
MW: 294.72 g/mol
MDL Number: MFCD09729058
CAS Number: 953747-30-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR