N-(4-Amino-2-methylphenyl)-2-(3-fluorophenoxy)acetamide

Supplier: Matrix Scientific

049036-500MG 049036-2.5G
101886-048EA 364.62 USD
101886-048 101886-050
N-(4-Amino-2-methylphenyl)-2-(3-fluorophenoxy)acetamide
N-(4-Amino-2-methylphenyl)-2-(3-fluorophenoxy)acetamide

Hazard: Irritant

Formula: C₁₅H₁₅FN₂O₂
MW: 274.3 g/mol
MDL Number: MFCD09735169
CAS Number: 953904-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR