N-(3-Amino-4-chlorophenyl)-2-(3-fluorophenoxy)acetamide
Supplier: Matrix Scientific
049034-500MG
049034-2.5G
101886-040EA
364.62
USD
101886-040
101886-042
N-(3-Amino-4-chlorophenyl)-2-(3-fluorophenoxy)acetamide
N-(3-Amino-4-chlorophenyl)-2-(3-fluorophenoxy)acetamide
Hazard: Irritant
Formula:
C₁₄H₁₂ClFN₂O₂ MW: 294.72 g/mol |
MDL Number:
MFCD09741487 CAS Number: 954575-59-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...