N-(5-Amino-2-methoxyphenyl)-2-(3-fluorophenoxy)acetamide
Supplier: Matrix Scientific
049032-500MG
049032-2.5G
101886-032EA
364.62
USD
101886-032
101886-034
N-(5-Amino-2-methoxyphenyl)-2-(3-fluorophenoxy)acetamide
N-(5-Amino-2-methoxyphenyl)-2-(3-fluorophenoxy)acetamide
Hazard: Irritant
Formula:
C₁₅H₁₅FN₂O₃ MW: 290.3 g/mol |
MDL Number:
MFCD09735962 CAS Number: 953896-36-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...