N-(3-Amino-4-fluorophenyl)-2-(3-fluorophenoxy)acetamide
Supplier: Matrix Scientific
049031-500MG
049031-2.5G
101886-028EA
364.62
USD
101886-028
101886-030
N-(3-Amino-4-fluorophenyl)-2-(3-fluorophenoxy)acetamide
N-(3-Amino-4-fluorophenyl)-2-(3-fluorophenoxy)acetamide
Hazard: Irritant
Formula:
C₁₄H₁₂F₂N₂O₂ MW: 278.26 g/mol |
MDL Number:
MFCD09736870 CAS Number: 954263-27-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...