N-(5-Amino-2-fluorophenyl)-2-(3-fluorophenoxy)acetamide
Supplier: Matrix Scientific
049030-500MG
049030-2.5G
101886-024EA
364.62
USD
101886-024
101886-026
N-(5-Amino-2-fluorophenyl)-2-(3-fluorophenoxy)acetamide
N-(5-Amino-2-fluorophenyl)-2-(3-fluorophenoxy)acetamide
Hazard: Irritant
Formula:
C₁₄H₁₂F₂N₂O₂ MW: 278.26 g/mol |
MDL Number:
MFCD09732125 CAS Number: 953754-32-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...