N-(4-Amino-2-methoxyphenyl)tetrahydro-2-furancarboxamide hydrochloride

Supplier: Matrix Scientific

049019-500MG 049019-2.5G
101885-980EA 166.15 USD
101885-980 101885-982
N-(4-Amino-2-methoxyphenyl)tetrahydro-2-furancarboxamide hydrochloride
N-(4-Amino-2-methoxyphenyl)tetrahydro-2-furancarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂O₃
MDL Number: MFCD28023815
CAS Number: 1706429-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR