N-(4-Aminophenyl)tetrahydro-2-furancarboxamide

Supplier: Matrix Scientific

049017-500MG 049017-2.5G
101885-972EA 364.62 USD
101885-972 101885-974
N-(4-Aminophenyl)tetrahydro-2-furancarboxamide
N-(4-Aminophenyl)tetrahydro-2-furancarboxamide

Hazard: Irritant

Formula: C₁₁H₁₄N₂O₂
MDL Number: MFCD09045362
CAS Number: 926223-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR