N-(3-Amino-4-chlorophenyl)tetrahydro-2-furancarboxamide

Supplier: Matrix Scientific

049016-500MG 049016-2.5G
101885-968EA 364.62 USD
101885-968 101885-970
N-(3-Amino-4-chlorophenyl)tetrahydro-2-furancarboxamide
N-(3-Amino-4-chlorophenyl)tetrahydro-2-furancarboxamide

Hazard: Irritant

Formula: C₁₁H₁₃ClN₂O₂
MDL Number: MFCD09050880
CAS Number: 926231-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR