N-(5-Amino-2-chlorophenyl)tetrahydro-2-furancarboxamide
Supplier: Matrix Scientific
049015-500MG
049015-2.5G
101885-964EA
364.62
USD
101885-964
101885-966
N-(5-Amino-2-chlorophenyl)tetrahydro-2-furancarboxamide
N-(5-Amino-2-chlorophenyl)tetrahydro-2-furancarboxamide
Hazard: Irritant
Formula:
C₁₁H₁₃ClN₂O₂ |
MDL Number:
MFCD11568388 CAS Number: 1082156-62-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...