N-(3-Amino-4-fluorophenyl)tetrahydro-2-furancarboxamide

Supplier: Matrix Scientific

049013-500MG 049013-2.5G
101885-956EA 364.62 USD
101885-956 101885-958
N-(3-Amino-4-fluorophenyl)tetrahydro-2-furancarboxamide
N-(3-Amino-4-fluorophenyl)tetrahydro-2-furancarboxamide

Hazard: Irritant

Formula: C₁₁H₁₃FN₂O₂
MDL Number: MFCD09043903
CAS Number: 926261-94-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR